
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:126456-43-7
:
Formula:C₉H₁₁NO
MDL Number:MFCD00216655
:149.19 g/mol
Melting Pt:117…118 °C
Storage Temperature:Ambient
ADR:8,III
:GHS05
Related Information
Frequently Bought Together