
tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate 97%
Catalog # H29152.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
Specifications
About this item
CAS Number:325775-44-8
:
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD01861762
:186.25 g/mol
:GHS07
Storage Temperature:Ambient