

tert-Butoxybis(dimethylamino)methane 90%, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5815-08-7
:
Formula:C₉H₂₂N₂O
MDL Number:MFCD00042858
:174.288 g/mol
Boiling Pt:50…55 °C (15 mmHg)
Density:0.844
Flash Pt:41 °C (106 °F)
Storage Temperature:Ambient
EINECS:227-383-9
UN:1993
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together