
7-Bromo-2H-1,4-benzothiazin-3(4H)-one 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:90814-91-8
:
Formula:C₈H₆BrNOS
MDL Number:MFCD02660583
:244.11 g/mol
Melting Pt:210…214 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together