



2-(Ethoxycarbonyl)ethylzinc bromide 0.5 M in THF
Catalog # H26739.AD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mL
- Seq:2370
Specifications
About this item
CAS Number:193065-68-8
:
Formula:BrZnCH₂CH₂COOCH₂CH₃
MDL Number:MFCD00671976
:246.42 g/mol
Density:0.972
Flash Pt:–17 °C (1 °F)
UN:3399
ADR:4.3,II
:GHS02
:GHS05
:GHS08
:GHS07
Storage Temperature:Refrigerator