2-Methyl-3-phenyl-1-propene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3290-53-7
:
Formula:C₁₀H₁₂
MDL Number:MFCD00039815
:132.21 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together