Bisabolene mixture of isomers 96% (sum of isomers)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:495-62-5
:
Formula:C₁₅H₂₄
MDL Number:MFCD00129080
:204.36 g/mol
Density:0.89
Storage Temperature:Ambient
EINECS:207-805-8
Frequently Bought Together