
Polyethylene glycol monomethyl ether methanesulphonate, M.W. 1,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:175172-61-9
:
Formula:CH₃(OCH₂CH₂)nOSO₂CH₃
MDL Number:MFCD03100948
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together