Platinum : rhodium (70:30) (w/w) 30, wire for ISA type B standard grade thermocouple, Ø 0.33mm (0.013in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD00198186
Storage Temperature:Ambient
Related Information
Frequently Bought Together