Order Entry
Netherlands
ContactUsLinkComponent
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) (15% Pd)
Irritant
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) (15% Pd)
Catalog # 45511.03
CAS Number:  
Irritant
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) (15% Pd)
Catalog # 45511.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :708.08 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:887919-35-9
Formula:C₃₂H₅₆Cl₂N₂P₂Pd
MDL Number:MFCD09265123