Order Entry
Netherlands
ContactUsLinkComponent
Bis(tricyclohexylphosphine)palladium(II) dichloride (14.4% Pd)
Irritant
Bis(tricyclohexylphosphine)palladium(II) dichloride (14.4% Pd)
Catalog # 44844.03
CAS Number:  
Irritant
Bis(tricyclohexylphosphine)palladium(II) dichloride (14.4% Pd)
Catalog # 44844.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

 :
 :738.19 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:29934-17-6
Formula:C₃₆H₆₆Cl₂P₂Pd
MDL Number:MFCD00191830