Tebbe Reagent 0.5 M in toluene
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 67719-69-1 Molecular Formula: C13H18AlClTi Formula Weight: 284.62 Flash Point: 1°(34°F) Physical Form: Liquid Density: 0.927 Sensitivity: Air and Moisture Sensitive Appearance: Dark red-brown UN No.: UN3399 MDL No.: MFCD00151575
Synonyms: µ-Chloro-µ-methylenebis(cyclopentadienyl)titaniumdimethylaluminum, Bis(cyclopentadienyl-mu-chloro(dimethylaluminium-mu-methylenetitanium
- CAS Number:67719-69-1
- Formula:C₁₃H₁₈AlClTi
- MDL Number:MFCD00151575
- Molecular Weight:284.62 g/mol
- Merck Index:14,9091
- Density:0.927
- Flash Pt:1 °C (34 °F)
- Storage Temperature:Ambient
- UN:3399
- ADR:4.3,II
2 Options Available Below
Product Details & Documents
CAS No.: 67719-69-1 Molecular Formula: C13H18AlClTi Formula Weight: 284.62 Flash Point: 1°(34°F) Physical Form: Liquid Density: 0.927 Sensitivity: Air and Moisture Sensitive Appearance: Dark red-brown UN No.: UN3399 MDL No.: MFCD00151575
Synonyms: µ-Chloro-µ-methylenebis(cyclopentadienyl)titaniumdimethylaluminum, Bis(cyclopentadienyl-mu-chloro(dimethylaluminium-mu-methylenetitanium
- CAS Number:67719-69-1
- Formula:C₁₃H₁₈AlClTi
- MDL Number:MFCD00151575
- Molecular Weight:284.62 g/mol
- Merck Index:14,9091
- Density:0.927
- Flash Pt:1 °C (34 °F)
- Storage Temperature:Ambient
- UN:3399
- ADR:4.3,II



