
Tris(4,4,4-trifluoro-1-(2-thienyl)-1,3-butanediono)europium(III) hydrate
Catalog # 43345.02
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
:851.818 g/mol
Melting Pt:143…144 °C
EINECS:237-892-8
:GHS07
Storage Temperature:Ambient
CAS Number:14054-87-6
Formula:C₂₄H₁₇EuF₉O₇S₃
MDL Number:MFCD00041948