Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-Dichlorobis(triethylphosphine)platinum(II)
cis-Dichlorobis(triethylphosphine)platinum(II)
cis-Dichlorobis(triethylphosphine)platinum(II)
39284.ME
:
Thermo Fisher Scientific
:
cis-Dichlorobis(triethylphosphine)platinum(II)
39284.ME
:
Thermo Fisher Scientific
:
Pk:
500 mg
Seq:
2725
:
:
502.304 g/mol
:
139…142 °C
:
GHS08
:
GHS07
:
Ambient
:
15692-07-6
:
PtCl₂[P(C₂H₅)₃]₂
:
MFCD00058821