

Manganese(II) 2-ethylhexanoate 40% (w/w) in mineral spirits (6% Mn)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13434-24-7
:
Formula:C₁₆H₃₀MnO₄
MDL Number:MFCD00058698
:343.361 g/mol
Density:0.897
Flash Pt:40 °C (104 °F)
Storage Temperature:Ambient
UN:1268
ADR:3,III
:GHS08
:GHS02
Related Information
Frequently Bought Together