Potassium hexacyanoferrate(II) standardised solution 0.1 N in aqueous solution
Catalog # 35585.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:368.35 g/mol
Merck Index:14,7631
Storage Temperature:Ambient
CAS Number:13943-58-3
Formula:C₆FeK₄N₆
MDL Number:MFCD00167022