
Lithium thiocyanate hydrate
Catalog # 11349.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 g
Specifications
About this item
:
:65.02 g/mol (anhy)
EINECS:209-135-1
:GHS07
Storage Temperature:Ambient
CAS Number:123333-85-7
Formula:LiSCN·xH₂O
MDL Number:MFCD00149773