Anti-MTA2 Mouse Monoclonal Antibody [clone: F-9]
Catalog # SANTSC-55566
Supplier: Santa Cruz Biotechnology
Specifications
- Antibody type:Primary
- Antigen name:Metastasis Associated 1 Family Member 2
- Antigen symbol:MTA2
- Clonality:Monoclonal
- Clone:F-9
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Gene ID:9219
- Antigen synonyms:PID|MTA1L1
- Amino acid number:499 to 668
- Molecular weight:75 kDa
- Storage temperature:+4 °C
- Size:200 µg/ml
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: MTA2 (metastasis associated 1 family member 2)
Clonality: Monoclonal
Clone: F-9
Conjugation:
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat