Anti-CSNK1D mouse monoclonal antibody [clone: C-8]
SANTSC-55553
- Antibody type:Primary
- Antigen name:casein kinase 1 delta
- Antigen symbol:CSNK1D
- Clonality:Monoclonal
- Clone:C-8
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:Amino acids range 296 to 355
- Gene ID:1453
- Antigen synonyms:ASPS, HCKID, FASPS2, CKIdelta
- Amino acid number:296 to 355
- Molecular weight:49 kDa
- Storage temperature:+4 °C
- Size:200 µg/ml
- Pk:200 µG
Type: Primary
Antigen: CSNK1D (casein kinase 1 delta)
Clonality: Monoclonal
Clone: C-8
Conjugation:
Epitope: C-terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat