Order Entry
Netherlands
ContactUsLinkComponent
Anti-MMP2 Mouse Monoclonal Antibody [clone: 4D3]
Anti-MMP2 Mouse Monoclonal Antibody [clone: 4D3]
  SANTSC-53630
Anti-MMP2 Mouse Monoclonal Antibody [clone: 4D3]
  SANTSC-53630

 

  • Antibody type:
    Primary
  • Antigen name:
    matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase)
  • Antigen symbol:
    MMP2
  • Clonality:
    Monoclonal
  • Clone:
    4D3
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Amino acids range 557 to 569
  • Gene ID:
    4313
  • Antigen synonyms:
    MMP-2|MMP-II|MONA|CLG4A|CLG4|TBE-1
  • Amino acid number:
    557 to 569
  • Molecular weight:
    Pro-MMP-2: 72 kDa; Cleaved MMP-2: 63 kDa
  • Storage temperature:
    +4 °C
  • Size:
    200 µg/ml
  • Pk:
    200 µG

 

 

Type: Primary
Antigen: MMP2 (matrix metallopeptidase 2)
Clonality: Monoclonal
Clone: 4D3
Conjugation:
Epitope: Amino acids range 557 to 569
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rabbit, Rat