Anti-HLA-C Mouse Monoclonal Antibody [clone: H-5]
SANTSC-166134
- Antibody type:Primary
- Antigen name:major histocompatibility complex, class I, C
- Antigen symbol:HLA-C
- Clonality:Monoclonal
- Clone:H-5
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG3
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:3107
- Antigen synonyms:D6S204|HLC-C|HLA-JY3|PSORS1|HLAC
- Molecular weight:43 kDa
- Storage temperature:+4 °C
- Size:200 µg/ml
- Pk:200 µG
Type: Primary
Antigen: HLA-C (major histocompatibility complex, class I, C)
Clonality: Monoclonal
Clone: H-5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human