Goat IgG Isotype Control
Catalog # SANTSC-2028
Supplier: Santa Cruz Biotechnology
Specifications
- Antibody type:Control
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Storage temperature:+4 °C
- Size:200 µg/0,5 ml
- Pk:200 µG
Specifications
About this item
It can be used as negative control for Western blotting, immunoprecipitation and immunohistochemistry applications.
Type: Control
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: