Rabbit IgG Isotype Control (Agarose)
SANTSC-2345
- Antibody type:Control
- Conjugation:Agarose
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Storage temperature:+4 °C
- Size:250 µg/0,5 ml
- Pk:250 µG
It can be used as negative control for immunoprecipitation.
Type: Control
Antigen:
Clonality:
Clone:
Conjugation: Agarose
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: