Order Entry
Netherlands
ContactUsLinkComponent
Rabbit IgG Isotype Control
Rabbit IgG Isotype Control
Catalog # SANTSC-2027
Rabbit IgG Isotype Control
Catalog # SANTSC-2027

Specifications

  • Antibody type:
    Control
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage temperature:
    +4 °C
  • Size:
    200 µg/0,5 ml
  • Pk:
    200 µG

Specifications

About this item

It can be used as negative control for Western blotting, immunoprecipitation and immunohistochemistry applications.

Type: Control
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: