Anti-CTSK Mouse Monoclonal Antibody [clone: E-7]
Catalog # SANTSC-48353
Supplier: Santa Cruz Biotechnology
Specifications
- Antibody type:Primary
- Antigen name:cathepsin K
- Antigen symbol:CTSK
- Clonality:Monoclonal
- Clone:E-7
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG3
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:Amino acids range 191 to 240
- Gene ID:1513
- Antigen synonyms:CTSO2|CTS02|CTSO1|PYCD|CTSO|PKND
- Amino acid number:191 to 240
- Molecular weight:39 kDa
- Storage temperature:+4 °C
- Size:200 µg/ml
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: CTSK (cathepsin K)
Clonality: Monoclonal
Clone: E-7
Conjugation:
Epitope: Amino acids range 191 to 240
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse