Order Entry
Netherlands
ContactUsLinkComponent
Anti-c-Myc Antibody [clone: 9E10]
Anti-c-Myc Antibody [clone: 9E10]
Catalog # SANTSC-40
Anti-c-Myc Antibody [clone: 9E10]
Catalog # SANTSC-40

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    v-myc avian myelocytomatosis viral oncogene homolog
  • Antigen symbol:
    MYC
  • Clonality:
    Monoclonal
  • Clone:
    9E10
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Monkey,
    Human,
    Dog,
    Rat,
    Mouse,
    Cat
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    4609
  • Antigen synonyms:
    MRTL|bHLHe39|c-Myc|MYCC
  • Amino acid number:
    408 to 439
  • Molecular weight:
    67 kDa
  • Storage temperature:
    +4 °C
  • Size:
    200 µg/ml
  • Pk:
    200 µG

Specifications

About this item

Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: Monoclonal
Clone: 9E10
Conjugation:
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Cat, Dog, Human, Monkey, Mouse, Rat