Anti-CD3 Mouse Monoclonal Antibody [clone: PC3/188A]
SANTSC-20047
- Antibody type:Primary
- Antigen symbol:CD3
- Clonality:Monoclonal
- Clone:PC3/188A
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Amino acid number:156 to 168
- Molecular weight:25 kDa
- Storage temperature:+4 °C
- Size:200 µg/ml
- Pk:1 KIT
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: PC3/188A
Conjugation: Conjugated
Epitope: Amino acid range 156 to168
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse and Rat