Anti-Na+/K+-ATPase beta1 Mouse Monoclonal Antibody [clone: C464.8]
SANTSC-21713
- Antibody type:Primary
- Antigen name:ATPase Na+/K+ transporting subunit beta 1
- Antigen symbol:ATP1B1
- Clonality:Monoclonal
- Clone:C464.8
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Rabbit,Human,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:External domain of the β1 subunit
- Gene ID:481
- Antigen synonyms:ATP1B
- Molecular weight:40-60 kDa
- Storage temperature:+4 °C
- Size:200 µg/ml
- Pk:1 KIT
Type: Primary
Antigen: ATP1B1 (ATPase Na+/K+ transporting subunit beta 1)
Clonality: Monoclonal
Clone: C464.8
Conjugation:
Epitope: External domain of the β1 subunit
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse, Rat and Rabblt