Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: 4F10]
Catalog # SANTSC-69703
Supplier: Santa Cruz Biotechnology
Specifications
- Antibody type:Primary
- Antigen name:Heat shock protein 90kDa alpha family class A member 1
- Antigen symbol:HSP90AA1
- Clonality:Monoclonal
- Clone:4F10
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Gene ID:3320
- Antigen synonyms:HSPCA|Hsp89|EL52|HSP86|LAP-2|HSPCAL1|HSPN|Hsp90|HSPC1|HSP90N|HSPCAL4|LAP2|HEL-S-65p|HSP89A|HSP90A
- Molecular weight:90 kDa
- Storage temperature:+4 °C
- Size:50 µg/0,5 ml
- Pk:1
Specifications
About this item
Type: Primary
Antigen: HSP90AA1 (Heat shock protein 90kDa alpha family class A member 1)
Clonality: Monoclonal
Clone: 4F10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human