Anti-ATF2 mouse monoclonal antibody [clone: F2BR-1]
Catalog # SANTSC-242
Supplier: Santa Cruz Biotechnology
Specifications
- Type antilichaam:Primair
- antigeen symbool:ATF2
- Clonality:Monoklonaal
- Clone:F2BR-1
- Host:Mouse
- ImmunoFluorescentie:Yes
- ImmunoNeerslag:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitoop:Amino acid range 350 to 505
- Gen-ID:1386
- Antigeen synoniemen:TREB7|HB16|CREB2|CREB-2|CRE-BP1
- Aminozuurgetal:350 to 505
- Moleculair gewicht:70 kDa
- Opslagtemperatuur:+4 °C
- Maat:200 µg/ml
- VPE:200 µG
Specifications
About this item
Type: Primary
Antigen: ATF2 (activating transcription factor 2)
Clonality: Monoclonal
Clone: F2BR-1
Conjugation:
Epitope: Amino acid range 350 to 505
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse