Anti-HNRNPA2B1 Mouse Monoclonal Antibody [clone: DP3B3]
Catalog # SANTSC-32316
Supplier: Santa Cruz Biotechnology
Specifications
- Antibody type:Primary
- Antigen name:Heterogeneous nuclear ribonucleoprotein A2/B1
- Antigen symbol:HNRNPA2B1
- Clonality:Monoclonal
- Clone:DP3B3
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Pig,Human,Bovine,Dog,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:3181
- Antigen synonyms:HNRPA2B1|HNRPA2|HNRPB1|HNRNPA2|SNRPB1|IBMPFD2|RNPA2|HNRNPB1
- Molecular weight:36/38 kDa
- Storage temperature:+4 °C
- Size:200 µg/ml
- Pk:1 KIT
Specifications
About this item
Type: Primary
Antigen: HNRNPA2B1 (heterogeneous nuclear ribonucleoprotein A2/B1)
Clonality: Monoclonal
Clone: DP3B3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Dog, Human, Mouse, Pig