Cation exchange chromatography column, HiScreen™ Capto™ S ImpAct
Catalog # 17-3717-47
Supplier: Cytiva
Specifications
- Flow rate:1,2 - 2,3 ml/min
- Particle Size:50 µm
- Capacity:4,7 ml
- Description:HiScreen™ Capto™ S ImpAct
- Seq:01
- pH range:4 - 12
- Binding capacity:> 85 mg BSA/ml medium, > 90 mg lysozyme/ml medium, > 100 mg IgG/ ml medium
- Storage:4 to 30 °C, 20% Ethanol
- Column hardware:PP
- Bed dimensions:7,7×100 mm
- Flow velocity:100 - 300 cm/h
- Pk:1
Specifications
About this item
HiScreen™ Capto™ S ImpAct columns are prepacked with Capto™ S ImpAct cation exchange chromatography medium. The columns are an excellent choice for method optimisation and parameter screening.
- Small bed volume gives fast results and minimal sample/buffer consumption
- Easily connected in series to achieve 200 mm bed height
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
- Strong cation exchange BioProcess medium, designed for immediate purification and polishing of a wide range of biomolecules, especially monoclonal antibodies
- Flow rate of 1,2 to 2,3 ml/min