Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Latex gloves, Rotiprotect®
Latex gloves, Rotiprotect®
Marketsource
Leverancier:   Roth Carl
Latex gloves, Rotiprotect®
Leverancier:  Roth Carl
Marketsource
Certificates
2 Opties Hieronder Beschikbaar
Product Details & Documents
These disposable gloves can be used for medical examinations, laboratory work and food applications.
  • Powder-free
  • Maximum wearing comfort and optimum perspiration absorption
  • Intensive washing processes ensure gloves contain low levels of protein and irritants
  • Thiuram-free
  • Textured surface for secure grip
  • Synthetic inner coating makes gloves easy to put on
 : EN 455, EN 374; Cat. III; AQL 1,5
Food contact approved
Specifications for Product Family
Specificaties
  • Catalog No:
  • 112-0066
  • 112-0067
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex
  • Latex
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Finish:
  • Textured
  • Textured
  • Cuff style:
  • Beaded
  • Beaded
  • EN norm:
  • EN 455, EN 374
  • EN 455, EN 374
  • Food contact approved:
  • +
  • +
Specificaties
Product Family Options
Productinformatie
  • Glove size
    Colour
    Length
    Thickness
    Packed
    Beschikbaarheid
    Price
  • S
    Natural
    240 mm
    0,22 mm
    100/box
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 100 ST
    1 * 100 ST - 13,55 €
    13,55 €
    1 * 100 ST
       13,55 €
    Specificaties:
    Marketsource
    More
    Specifications
    Cat. No.
    112-0066
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex
    Powdered / powder-free
    Powder-free
    Finish
    Textured
    Cuff style
    Beaded
    EN norm
    EN 455, EN 374
    Food contact approved
    +
    Marketsource
  • M
    Natural
    240 mm
    0,22 mm
    100/box
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 100 ST
    1 * 100 ST - 13,55 €
    13,55 €
    1 * 100 ST
       13,55 €
    Specificaties:
    Marketsource
    More
    Specifications
    Cat. No.
    112-0067
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex
    Powdered / powder-free
    Powder-free
    Finish
    Textured
    Cuff style
    Beaded
    EN norm
    EN 455, EN 374
    Food contact approved
    +
    Marketsource