Order Entry
Netherlands
ContactUsLinkComponent
1,4-Bis(2-methylstyryl)benzene for synthesis, Sigma-Aldrich®
1,4-Bis(2-methylstyryl)benzene for synthesis, Sigma-Aldrich®
Catalog # 8.41875.0001
Supplier:  Merck
CAS Number:  
1,4-Bis(2-methylstyryl)benzene for synthesis, Sigma-Aldrich®
Catalog # 8.41875.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Melting Point:
    179 - 181 °C
  • Molar Mass:
    310.43 g/mol
  • Synonyms:
    Bis-MSB
  • Hill Formula:
    C24H22
  • HS Code:
    2902 90 00
  • Storage temperature:
    15...25 °C
  • Assay (HPLC, area%):
    ≥97% (a/a)
  • Identity (IR):
    passes test

Specification Test Results

About this item

 :
 :310.44 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:236-285-5
CAS Number:13280-61-0
Formula:C₂₄H₂₂
MDL Number:MFCD00008529