meso-Hydrobenzoin for synthesis, Sigma-Aldrich®
8.20526.0001
: Merck
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
- Solubility:(20 °C) slightly soluble
- Melting Point:128 - 131 °C
- Molar Mass:214.26 g/mol
- Synonyms:meso-Hydrobenzoin, meso-1,2-Diphenylethylene glycol
- Hill Formula:C14H14O2
- Chemical Formula:HOCH(C6H5)CH(C6H5)OH
- Storage temperature:+15...+25 °C
: meso-1,2-Diphenylethylene glycol, meso-1,2-Diphenyl-1,2-ethanediol, rel-(1R,2S)-1,2-Diphenylethane-1,2-diol
:
:214.26 g/mol
:M-Clarity™ quality level = MQ200
:Ambient
:207-758-3
:492-70-6
:C₁₄H₁₄O₂
:MFCD00004499