Anti-MAP2K3 Rabbit Polyclonal Antibody
Catalogus # BOSSBS-4123R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase Kinase 3
- Antigen symbol:MAP2K3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:5606
- Antigen synonyms:dual specificity mitogen-activated protein kinase kinase 3|SAPKK-2|MEK 3|MAP kinase kinase 3|MAPKK3|SAPK kinase 2|SAPKK2|PRKMK3|stress-activated protein kinase kinase 2|MKK3|MAPKK 3|MAPK/ERK kinase 3
- Modification:Thr222
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Dual specificity kinase. Is activated by cytokines and environmental stress in vivo. Catalyzes the concomitant phosphorylation of a threonine and a tyrosine residue in the MAP kinase p38.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: MAP2K3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat