Anti-MHC class I antigen B 15 Rabbit Polyclonal Antibody
Catalog # BOSSBS-4120R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:MHC class I Antigen B 15
- Antigen symbol:MHC class I Antigen B 15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Gene ID:3106
- Antigen synonyms:MHC class I antigen B 13|MHC class I antigen B 15.|HLA-B15,HLA class I histocompatibility antigen B 13 alpha chain precursor|HLA class I histocompatibility antigen B 15 alpha chain precursor
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
HLA B belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta 2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells.
WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: MHC class I antigen B 15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human