Order Entry
Netherlands
ContactUsLinkComponent
Anti-CALHM1 Rabbit Polyclonal Antibody
Anti-CALHM1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-11642R
Supplier:  Bioss
Anti-CALHM1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-11642R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Calcium Homeostasis Modulator 1
  • Antigen symbol:
    CALHM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    FAM26C|Protein FAM26C|CALHM1|Calcium homeostasis modulator protein 1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

CALHM1 is a 346 amino acid multi-pass endoplasmic reticulum membrane protein that belongs to the FAM26 family. CALHM1 co-localizes with GRP 78 to the endoplasmic reticulum. Predominantly expressed in adult brain, CALHM1 may be a pore-forming ion channel that controls cytosolic Ca2+ permeability and cytosolic Ca2+ concentration in the cell. It is suggested that CALHM1 regulates amyloid precursor protein proteolysis and aggregated amyloid-beta peptides levels in a Ca2+ dependent manner. CALHM1 homomultimerizes and shares strong sequence similarities with the selectivity filter of the NMDA receptor, which generates a large Ca2+ conductance across the plasma membrane. CALHM1 may be a potential factor involved in the pathogenesis of Alzheimer's disease.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CALHM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat