Trimming blades and handles, FEATHER®
Supplier: PFM Medical GmbH
Certificates
About this item
The carbon steel trimming blades are special blades for cutting tissue samples without causing deformations. The trimming blade handles are made from heat-resistant plastic and due to their ergonomic design, they are particularly easy to grip.
- Super sharp blades
- Securely-fitted blade can be easily attached and removed