Order Entry
Netherlands
ContactUsLinkComponent
Sterilsop® SX removable heat sterilization bags
Sterilsop® SX removable heat sterilization bags
Catalog # HART6555051
Supplier:  HARTMANN PAUL
Sterilsop® SX removable heat sterilization bags
Catalog # HART6555051
Supplier:  HARTMANN PAUL
Specifications
  • L×W:
    550×320 mm
  • Seq:
    08
  • Pk:
    300
Specifications
About this item
Heat-sealable sterilization packages specially designed for wet heat sterilisation. Sterilsop® SX sachets are peelable flat bags and are intended for use as sterile barrier systems for the packaging of medical linen, medical and surgical equipment, dressing supplies.
  • Permeability to the sterilising agent
  • Maintenance of sterility of content
  • Aseptic extraction of sterilised material
  • One side paper with sterilisation indicators
  • A complex pink transparent polyester / polypropylene face
  • Free of latex, phthalates and rosin
Sterilsop® SX is a Class I device.
Shelf life: five years
Certifications: Complies with EN ISO 11607-1 and EN 868-5 standards.