Order Entry
Netherlands
ContactUsLinkComponent
 
VWR®, Digital Timer, One Channel
  609-0210
undefined
VWR®, Digital Timer, One Channel
  609-0210

 

  • Colour:
    Black
  • Description:
    Digital timer, single channel
  • W×D×H:
    63,5×12,7×63,5 mm
  • Accuracy:
    ±0,01 %
  • Pk:
    1

 

 

Counts down from 99 minutes, 59 seconds in second increments.

  • Large digits (19 mm) are readable from a distance of 6 metres
  • Three large keys makes it easy to use
  • Alarm sounds for 30 seconds and then silences automatically
  • Memory resets display to last programmed time for quick repeat of frequent, routine tests
 : Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
 : Supplied with magnet, flip-out stand, clip, battery, and certificate.