Anti-IDH2 Antibody
BSBTPB9602
- Antibody type:Primary
- Antigen name:Isocitrate dehydrogenase 2
- Antigen symbol:IDH2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3418
- Antigen synonyms:IDHM|D2HGA2|mNADP-IDH|IDH|ICD-M|IDPM|IDP
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Isocitrate dehydrogenase [NADP], mitochondrial(IDH2) detection. Tested with WB, IHC-P in Human; Mouse;Rat.
Type: Primary
Antigen: IDH2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat