Peqlab peqGREEN, DNA/RNA Dye
Supplier: VWR Peqlab
Certificates
About this item
peqGREEN dye is a safe alternative to ethidium bromide for staining of dsDNA, ssDNA and RNA.
- Direct addition to gel solution or staining after electrophoresis
- Excitation with UV or blue light
Suitable for up to 400 gels.
Frequently Bought Together