Gas trapping systems for LC/MS, Super Clean®
Catalog # REST22062
Supplier: Restek
Specifications
- For:LC-MS
- Description:Super Clean® gas trapping system, includes baseplate and 2 hydrocarbon filters
- Gases purified:Non-corrosive gases
- Outlet purity:99,9999%
- Filter:Hydrocarbon
- Max. operating pressure:11 bar
- Max. flow rate:20 l/min
- Pk:1 KIT
Specifications
About this item
The Super Clean® gas trapping system purifies nitrogen and is ideal for use in LC-MS systems. The two position base plate (installed in the gas line; standard base plate inlet/outlet fittings accept ¹/₄" tubing) allows cartridges to be exchanged without introducing impurities into the system. To meet the high flow needs of the LC-MS system, the activated charcoal filled cartridges are positioned and connected in parallel.
- Up to 20 L of hydrocarbon free nitrogen per minute
- Spring-loaded check valves allow cartridge replacement without contamination hazard
- Estimated filter lifetime of 3 to 6 months