Order Entry
Netherlands
ContactUsLinkComponent
Guaiacol detection kit
Guaiacol detection kit
Catalog # 710-0751
Supplier:  DOEHLER GROUP
CAS Number:  
undefined
Guaiacol detection kit
Catalog # 710-0751
Supplier:  DOEHLER GROUP
CAS Number:  
Specifications
  • Pack type:
    Bottle
  • Pk:
    40 Tests
  • Standards:
    IFU Method No. 12
  • Description:
    Guaiacol-detection- kit
  • Filling:
    -
Specifications
About this item
Enzymatic test kit for the detection of off-flavour forming Alicyclobacillus. Guaiacol Detection Kit is a reliable and sensitive enzymatic detection of Guaiacol-forming Alicyclobacilli. When Guaiacol-forming bacteria are present, the transparent to slightly yellow Guaiacol is transformed into the clearly-visible, brownish complex Tetraguaiacol with the enzyme peroxidase from the Detection Kit.
  • Comprehensive Detection Kit including BAT Broth in tubes and Guaiacol Standard Solution
  • Simple visual analysis without additional technical equipment