Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CDH1 Antibody Pair
  ABNOH00000999-AP61
 :  Abnova
Anti-CDH1 Antibody Pair
  ABNOH00000999-AP61
 :  Abnova

 

  • Antigen name:
    Cadherin 1
  • Antigen symbol:
    CDH1
  • Host:
    Multiple
  • Reactivity:
    Human
  • Gene ID:
    999
  • Antigen synonyms:
    CD324|epithelial Cadherin|Cadherin-E|ECAD|Arc-1|CAM 120/80|cell-CAM 120/80|E-Cadherin|uvomorulin|Cadherin 1|Cadherin-1|E-Cadherin epithelial|calcium-dependent adhesion protein|epithelial|LCAM|CDHE|UVO
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Set comes with matched Antibody Pair to detect and quantify protein level of human CDH1.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-CDH1, IgG1 kappa (100 µg); 2. Detection Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-CDH1 (50 µg);

Type:
Antigen: CDH1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human