Order Entry
Netherlands
ContactUsLinkComponent
Anti-PYCRL Mouse Monoclonal Antibody
Anti-PYCRL Mouse Monoclonal Antibody
  ABNOH00065263-PW3
 :  Abnova
Anti-PYCRL Mouse Monoclonal Antibody
  ABNOH00065263-PW3
 :  Abnova

 

  • Antigen name:
    Pyrroline-5-carboxylate reductase-like
  • Antigen symbol:
    PYCRL
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    65263
  • Antigen synonyms:
    PYCR3
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-PYCRL (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-PYCRL (50 ?g)

Type:
Antigen: PYCRL
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human