Order Entry
Netherlands
ContactUsLinkComponent
Glass and stainless steel cleaner, TASKI sprint 200
Glass and stainless steel cleaner, TASKI sprint 200
Catalog # DIVY7512859
Supplier:  Diversey
undefined
Glass and stainless steel cleaner, TASKI sprint 200
Catalog # DIVY7512859
Supplier:  Diversey
Specifications
  • Description:
    TASKI sprint 200, hard surface and glass cleaner
  • Packed:
    1 litre bottle
  • Pk:
    6 Bottle
Specifications
About this item
Solvent-based formulation that gives low residue and fast buff response to give streak-free finish on all surfaces including glass and stainless steel.
  • Multi-purpose for surfaces and floors
  • Suitable for use with trigger spray or bucket
  • Pleasant fragrance with ONT (Odour Neutralising Technology)