Volumetric flasks, Class A, USP, glass, clear, BLAUBRAND®, NS plastic
Catalog # BRNDUSP37249
Supplier: Brand
Specifications
- NS:12/21
- Capacity:100 ml
- Seq:06
- Suppl. to DIN:√
- Pk:1
- Tolerance:0,10 ± ml
Specifications
About this item
Borosilicate glass 3.3, clear with PP stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
Certifications: DIN EN ISO 1042. Conformity certified (DE-M marking).
Delivery: Supplied with a batch number, batch certificate and with USP individual certificate.