QC microorganisms for culture purposes and quality control applications, LYFO DISK®
Catalog # 0245L
Supplier: MICROBIOLOGICS
CAS Number:
Specifications
- Pk:6
- Description:Aspergillus brasiliensis ATCC® 16888
- Seq:245
Specifications
About this item
LYFO DISK® is a lyophilised pellet containing a single strain of a QC microorganism and is the most economical reference stock culture option. LYFO DISK® is known for its ease-of-use; simply re-hydrate the pellet and inoculate. LYFO DISK® is available in a re-sealable vial containing 6 pellets of a single QC microorganism strain.
- Product warranty
- Does not require frozen storage conditions
- Traceable to reference culture